4,4'-Di-tert-butylbiphenyl 99+%

Synonyms: 4,4'-Di-tert-butyl-1,1'-biphenyl1-tert-butyl-4-(4-tert-butylphenyl)benzene

AC29188-0050
200010-122EA 45.9 USD
200010-122
4,4'-Di-tert-butylbiphenyl 99+%
4,4'-Di-tert-butylbiphenyl
Formula: C₂₀H₂₆
MW: 266.43 g/mol
Boiling Pt: 190…192 °C (13 mmHg)
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00008834
CAS Number: 1625-91-8
EINECS: 216-615-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR