Phenylsilane 97+%

Danger

Synonyms: Silylbenzene

AC29169-1000 AC29169-0250
103527-320EA 716 USD
103527-320 200007-346
Phenylsilane 97+%
Phenylsilane
Formula: C₆H₈Si
MW: 108.22 g/mol
Boiling Pt: 120 °C (760 mmHg)
Density: 0.87 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00013478
CAS Number: 694-53-1
EINECS: 211-772-5
UN: 3399
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR