Bis(4-chlorophenyl) sulfone 97%

Warning

Synonyms: 4,4'-Dichloro diphenyl sulphone4-Chlorophenyl sulfone4,4'-Dichloro diphenyl sulfone4,4'-Sulfonylbis(chlorobenzene)4-Chlorophenyl sulphone4,4'-sulphonylbis(chlorobenzene)

AC29118-0250 AC29118-2500 AC29118-0010
200047-646EA 123 USD
200047-646 200047-648 200029-508
Bis(4-chlorophenyl) sulfone 97%
Bis(4-chlorophenyl) sulphone
Formula: C₁₂H₈Cl₂O₂S
MW: 287.17 g/mol
Boiling Pt: 397 °C
Melting Pt: 146…151 °C
Flash Pt: 233 °C
MDL Number: MFCD00000619
CAS Number: 80-07-9
EINECS: 201-247-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR