Octanophenone 99+%

Synonyms: n-Octanophenone

AC27936-1000 AC27936-0050 AC27936-0250
103512-046EA 329 USD
103512-046 200050-324 200044-310
Octanophenone 99+%
Octanophenone
Formula: C₆H₅C(O)(CH₂)₆CH₃
MW: 204.31 g/mol
Boiling Pt: 285…290 °C (760 mmHg)
Melting Pt: 21.5…23.5 °C
Density: 0.93 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003554
CAS Number: 1674-37-9
EINECS: 216-817-2
https://us.vwr.com/stibo/web/std.lang.all/22/80/762280.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR