Heptanophenone 98+%

AC27921-0050 AC27921-0250
200049-618EA 53.5 USD
200049-618 200043-624
Heptanophenone 98+%
Heptanophenone
Formula: C₆H₅C(O)(CH₂)₅CH₃
MW: 190.29 g/mol
Boiling Pt: 155 °C (15 mmHg)
Melting Pt: 17 °C
Density: 0.94 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00009539
CAS Number: 1671-75-6
EINECS: 216-802-0
https://us.vwr.com/stibo/web/std.lang.all/22/57/762257.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR