α,3,4-Trichlorotoluene 98+%

Danger

Synonyms: 1,2-Dichloro-4-(chloromethyl)benzene1-(Chloromethyl)-3,4-dichlorobenzene3,4-Dichlorobenzyl chloride

AC27905-0250 AC27905-5000 AC27905-1000
103532-088EA 237 USD
103532-088 200047-230 200016-836
α,3,4-Trichlorotoluene 98+%
α,3,4-Trichlorotoluene
Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 122…124 °C (14 mmHg)
Melting Pt: –3 °C
Density: 1.41 g/cm³
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00000906
CAS Number: 102-47-6
EINECS: 203-033-0
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR