2,5-Dibromo-p-xylene 98+%

Warning

Synonyms: 2,5-Dibromo-1,4-dimethylbenzene1,4-Dibromo-2,5-dimethylbenzene

AC27901-2500 AC27901-0500
103509-838EA 350 USD
103509-838 200033-776
2,5-Dibromo-p-xylene 98+%
2,5-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00000091
CAS Number: 1074-24-4
EINECS: 214-038-2
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR