α-Bromo-2-chlorotoulene 97%

Danger

Synonyms: 2-Chlorobenzyl bromide1-(Bromomethyl)-2-chlorobenzene

AC27893-0050 AC27893-1000 AC27893-0250
200016-502EA 118 USD
200016-502 103532-086 200022-566
α-Bromo-2-chlorotoulene 97%
α-Bromo-2-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 103…104 °C (10 mmHg)
Density: 1.586 g/cm³
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00000566
CAS Number: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR