Xenon difluoride 99.5+%

Danger

Synonyms: Xenon(II) fluoride

AC27855-0050
103527-284EA 454 USD
103527-284
Xenon difluoride 99.5+%
Xenon difluoride
Formula: F₂Xe
MW: 169.29 g/mol
Melting Pt: 129 °C
Storage Temperature: Ambient
MDL Number: MFCD00040538
CAS Number: 13709-36-9
EINECS: 237-251-2
UN: 3085
ADR: 5.1,II
Merck Index: 14,10072

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR