3-Chloro-4-fluorophenol 99%

Warning

AC27720-0250 AC27720-0050
200041-498EA 87.7 USD
200041-498 200041-500
3-Chloro-4-fluorophenol 99%
3-Chloro-4-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 104 °C (11 mmHg)
Melting Pt: 39…44 °C
Flash Pt: 108 °C
MDL Number: MFCD00002257
CAS Number: 2613-23-2
EINECS: 220-042-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR