3'-Chloropropiophenone 98%

Warning

Synonyms: 1-(3-Chlorophenyl)propan-1-onem-Chloropropiophenone

AC27658-2500 AC27658-0250
200009-898EA 23.9 USD
200009-898 200035-330
3'-Chloropropiophenone 98%
3'-Chloropropiophenone
Formula: C₉H₉ClO
MW: 168.62 g/mol
Boiling Pt: 124 °C (14 mmHg)
Melting Pt: 43…47 °C
Flash Pt: >110 °C
MDL Number: MFCD00009925
CAS Number: 34841-35-5
EINECS: 252-242-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR