5-Nitro-o-toluic acid 99+%

Warning

AC27657-0050 AC27657-0250 AC27657-0010
103464-806EA 22 USD
103464-806 200003-000 103464-808
5-Nitro-o-toluic acid 99+%
5-Nitro-o-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00007371
CAS Number: 1975-52-6
EINECS: 217-829-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR