Xanthydrol 97+%

Warning

Synonyms: 9-Hydroxyxanthene9H-Xanthen-9-ol9-Xanthenol

AC27152-0050 AC27152-0250
200010-884EA 50.2 USD
200010-884 200017-946
Xanthydrol 97+%
Xanthydrol
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Boiling Pt: 338 °C (1013 hPa)
Melting Pt: 124…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005057
CAS Number: 90-46-0
EINECS: 201-996-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR