3-Amino-4-hydroxybenzoic acid 98%

Warning

AC27149-0250 AC27149-0050
200023-146EA 39.71 USD
200023-146 200023-144
3-Amino-4-hydroxybenzoic acid 98%
3-Amino-4-hydroxybenzoic acid
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 198…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR