5-Fluoro-1-indanone 98%

Warning

Synonyms: 5-Fluoro-2,3-dihydro-1H-inden-1-one

AC27111-2500
200048-198EA 26.2 USD
200048-198
5-Fluoro-1-indanone 98%
5-Fluoro-1-indanone
Formula: C₉H₇FO
MW: 150.15 g/mol
Boiling Pt: 92…93 °C (3 mmHg)
Melting Pt: 35…39 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00041031
CAS Number: 700-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR