β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate 95+%, reduced

Synonyms: beta-Nicotinamide adenine dinucleotide disodium salt hydratebeta-NADH disodium salt hydrateβ-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate

AC27110-2500 AC27110-0050 AC27110-0250 AC27110-0010
103518-698EA 83.09 USD
103518-698 200002-972 103518-694 103518-696
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form) hydrate 95+%, reduced
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate
Formula: C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
MW: 727.43 g/mol
Melting Pt: 140…142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03791273
CAS Number: 1949720-50-6
EINECS: 210-123-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR