Sodium carbonate decahydrate ≥99%, Extra Pure

Supplier: ACROS ORGANICS MS

Warning

AC27107-1000 AC27107-0025 AC27107-0100
103518-692EA 15.44 USD InStock
103518-692 200032-816 103518-690
Sodium carbonate decahydrate ≥99%, Extra Pure
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
MW: 286.14 g/mol
Melting Pt: 33 °C
Density: 1.46 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1
EINECS: 207-838-8

Order Now

Specification Test Results

Appearance White Crystals
Titration with HCl ≥99.0 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤30 ppm
Aluminium (Al) ≤10 ppm
Iron (Fe) ≤4 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤50 ppm
Silicon dioxide (SiO2) ≤30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR