4-Bromo-1,2-dichlorobenzene 98+%

Warning

Synonyms: 1-Bromo-3,4-dichlorobenzene

AC26981-0250 AC26981-1000
200021-768EA 217 USD
200021-768 200021-770
4-Bromo-1,2-dichlorobenzene 98+%
4-Bromo-1,2-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 140…142 °C (30 mmHg)
Melting Pt: 23…24 °C
MDL Number: MFCD00040849
CAS Number: 18282-59-2
EINECS: 242-160-6
UN: 2810

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR