1-(2-Fluorophenyl)piperazine 97%

Danger

AC26869-1000 AC26869-0100
200039-838EA 198 USD
200039-838 200015-396
1-(2-Fluorophenyl)piperazine 97%
1-(2-Fluorophenyl)piperazine
Formula: C₁₀H₁₃FN₂
MW: 180.23 g/mol
Boiling Pt: 93 °C (0.5 mmHg)
Density: 1.141 g/cm³
MDL Number: MFCD00040726
CAS Number: 1011-15-0
EINECS: 213-780-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR