Thallium(I) nitrate 99.5%

Danger

AC26559-1000
103527-202EA 232 USD
103527-202
Thallium(I) nitrate 99.5%
Thallium (I) nitrate
Formula: NO₃Tl
MW: 266.39 g/mol
Boiling Pt: 433 °C (760 mmHg)
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR