(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99+%

Warning

Synonyms: (R)-BINAP(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl(R)-(+)-BINAP(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene

AC26553-2500 AC26553-0010 AC26553-0050
200033-390EA 36 USD
200033-390 200033-392 200027-350
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 99+%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…242 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR