3,4-Dibromothiophene 97%

Warning

AC26452-0010 AC26452-0250 AC26452-0050
200002-902EA 21.5 USD
200002-902 103532-026 103532-028
3,4-Dibromothiophene 97%
3,4-Dibromothiophene
Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221…222 °C (760 mmHg)
Melting Pt: 4…5 °C
Density: 2.13 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR