1-Chloroadamantane 98%

Warning

AC26424-0100 AC26424-2500 AC26424-0500
103511-442EA 943 USD
103511-442 200021-798 200008-652
1-Chloroadamantane 98%
1-Chloroadamantane
Formula: C₁₀H₁₅Cl
MW: 170.68 g/mol
Melting Pt: 163…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00075627
CAS Number: 935-56-8
EINECS: 213-305-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR