Cyclopropylamine 98%

Danger

AC26368-1000 AC26368-0250 AC26368-5000 AC26368-0025
200055-396EA 219 USD
200055-396 103527-180 103527-182 103527-184
Cyclopropylamine 98%
Cyclopropylamine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 49…50 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.824 g/cm³
Flash Pt: –25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR