2,5-Dibromotoluene 98%

Warning

Synonyms: 1,4-Dibromo-2-methylbenzene

AC26348-0050 AC26348-0250
200016-138EA 99.1 USD
200016-138 200016-136
2,5-Dibromotoluene 98%
2,5-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 135…136 °C (35 mmHg)
Melting Pt: 5…6 °C
Density: 1.81 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00000090
CAS Number: 615-59-8
EINECS: 210-437-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR