3-Thiophenecarbonitrile 94%

Warning

Synonyms: Thiophene-3-carbonitrile3-Thienyl cyanide3-Cyanothiophene

AC26316-0010
200029-446EA 41.7 USD
200029-446
3-Thiophenecarbonitrile 94%
3-Thiophenecarbonitrile
Formula: C₅H₃NS
MW: 109.15 g/mol
Boiling Pt: 82 °C (10 mmHg)
MDL Number: MFCD00151852
CAS Number: 1641-09-4
EINECS: 216-687-7
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR