3-Methoxy-3-methyl-1-butanol 99%
|
Warning
|
Synonyms:
MMB
, 3-Methoxy-3-methylbutan-1-ol
, Solvant 39
, 3-Methyl-3-methoxybutanol
AC25975-2500
AC25975-0010
200023-376EA
36
USD
200023-376
103465-392
3-Methoxy-3-methyl-1-butanol 99%
3-Methoxy-3-methyl-1-butanol
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Boiling Pt: 174 °C (760 mmHg) Melting Pt: –50 °C Density: 0.926 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044771 CAS Number: 56539-66-3 EINECS: 260-252-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...