Butyl butyrate 98%

Warning

Synonyms: Butyric acid butyl ester

AC25959-0010 AC25959-0050 AC25959-1000
200048-724EA 16.4 USD
200048-724 200030-628 200036-720
Butyl butyrate 98%
Butyl butyrate
Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –92 °C
Density: 0.8675 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 1993
ADR: 3,III
Merck Index: 13,01555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR