Cyclopropanemethylamine 97%

Danger

Synonyms: (Aminomethyl)cyclopropane(Cyclopropylmethyl)amineCyclopropylmethylamine

AC25872-0500 AC25872-0050 AC25872-0100
200011-188EA 210 USD
200011-188 103527-160 200011-190
Cyclopropanemethylamine 97%
Cyclopropanemethylamine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 84…85 °C (760 mmHg)
Density: 0.82 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00037147
CAS Number: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR