Copper (II) ethylenediamine complex 1 M in water

Danger

Synonyms: CupriethylendiamineBis(ethylenediamine)copper dihydroxideCopper(II)-ethylenediame complex

AC25855-0025 AC25855-0010
103527-154EA 133.85 USD
103527-154 200006-108
Copper (II) ethylenediamine complex 1 M in water
Copper (II) ethylenediamine
Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MW: 217.76 g/mol
Density: 1.1
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00274628
CAS Number: 14552-35-3
UN: 1761
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR