3-Methoxypropiononitrile 98%

Warning

Synonyms: 3-Methoxypropanenitrile3-Methoxypropionitrile

AC25744-5000 AC25744-0025 AC25744-1000
103511-902EA 267 USD
103511-902 200010-010 103511-904
3-Methoxypropiononitrile 98%
3-Methoxypropiononitrile
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 164…165 °C (760 mmHg)
Melting Pt: –62.9 °C
Density: 0.938 g/cm³
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR