1-Chlorododecane 99%

Warning

Synonyms: Lauryl chlorideDodecylchloride

AC25710-2500 AC25710-0010
200012-974EA 35.9 USD
200012-974 103531-994
1-Chlorododecane 99%
1-Chlorododecane
Formula: C₁₂H₂₅Cl
MW: 204.78 g/mol
Boiling Pt: 260 °C (760 mmHg)
Melting Pt: –9.3 °C
Density: 0.87 g/cm³
Flash Pt: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD00013688
CAS Number: 112-52-7
EINECS: 203-981-5
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR