3-Cyclohexyl-1-propanol 99%

Synonyms: 3-Cyclohexylpropan-1-ol3-Hydroxypropylcyclohexane

AC25588-0050
200029-268EA 73.9 USD
200029-268
3-Cyclohexyl-1-propanol 99%
3-Cyclohexyl-1-propanol
Formula: C₉H₁₈O
MW: 142.24 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.91 g/cm³
Flash Pt: 116 °C
MDL Number: MFCD00001528
CAS Number: 1124-63-6
EINECS: 214-401-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR