4-Methylvaleric acid 99%

Danger

Synonyms: Isocaproic acid4-Methylpentanoic acid

AC25551-0050 AC25551-0250
200017-646EA 102 USD
200017-646 200017-644
4-Methylvaleric acid 99%
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.922 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0
UN: 2922

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR