Disodium hydrogen citrate sesquihydrate 99%, pure
Synonyms:
Sodium hydrogencitrate sesquihydrate
, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate
, Sodium citrate dibasic sesquihydrate
, Citric acid disodium salt sesquihydrate
AC25024-0025
AC25024-2500
103517-758EA
194
USD
103517-758
103517-760
Disodium hydrogen citrate sesquihydrate 99%, pure
di-Sodium hydrogen citrate sesquihydrate
Formula:
C₆H₆Na₂O₇·1.5H₂O MW: 263.11 g/mol Melting Pt: 149 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00150445 CAS Number: 6132-05-4 EINECS: 242-734-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...