3-Iodophenol 99%

Warning

AC24970-0050 AC24970-0250
103511-878EA 30.2 USD
103511-878 200009-994
3-Iodophenol 99%
3-Iodophenol
Formula: C₆H₅IO
MW: 220.01 g/mol
Melting Pt: 38…43 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002261
CAS Number: 626-02-8
EINECS: 210-923-2
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR