Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Difluoroanisole ≥98%

Supplier: ACROS ORGANICS MS

Danger

AC24955-0050
200041-354EA 25.2 USD InStock
200041-354
3,4-Difluoroanisole ≥98%
3,4-Difluoroanisole


Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.23 g/cm³ (20 °C)
MDL Number: MFCD00010730
CAS Number: 115144-40-6
EINECS: 000-000-0
UN: 1993
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4675 to 1.4695 (20°C, 589 nm)
GC ≥98.0 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty