1-Bromo-3-iodobenzene 98+%

Warning

Synonyms: 3-Bromoiodobenzene

AC24942-0100 AC24942-0500
103511-868EA 217 USD
103511-868 200015-710
1-Bromo-3-iodobenzene 98+%
1-Bromo-3-iodobenzene
Formula: C₆H₄BrI
MW: 282.91 g/mol
Boiling Pt: 120 °C (18 mmHg)
Melting Pt: –9.3 °C
Density: 2.21 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001043
CAS Number: 591-18-4
EINECS: 209-703-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR