(2S,3S)-(+)-2,3-Butanediol 99%

Synonyms: L(+)-2,3-Butanediol(S,S)-(+)-2,3-Dihydroxy butane(2S,3S)-Butane-2,3-diol(S,S)-(+)-2,3-Butanediol(S,S)-(+)-2,3-Butylene glykol(2S,3S)-(+)-2,3-Dihydroxy butane(2S,3S)-(+)-2,3-Butylene glykol

AC24773-5000
200033-312EA 84.4 USD
200033-312
(2S,3S)-(+)-2,3-Butanediol 99%
(2S,3S)-(+)-2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 179…182 °C (760 mmHg)
Density: 0.98 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00063648
CAS Number: 19132-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR