2-Methylcyclopentanone ≥99%

Supplier: ACROS ORGANICS MS

Warning

AC24766-0050 AC24766-0250
200022-826EA 128.52 USD
200022-826 103527-100
2-Methylcyclopentanone ≥99%
2-Methylcyclopentanone
Formula: C₆H₁₀O
MW: 98.15 g/mol
Boiling Pt: 139…140 °C
Melting Pt: -75 °C
Density: 0.917 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00001414
CAS Number: 1120-72-5
EINECS: 214-318-4
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR