5-Chloro-2-nitroaniline 97%

Danger

AC24695-0500 AC24695-0050
200005-560EA 159 USD
200005-560 200030-066
5-Chloro-2-nitroaniline 97%
5-Chloro-2-nitroaniline
Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 126…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS Number: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR