5-Nitroanthranilic acid 98%

Warning

Synonyms: 2-Amino-5-nitrobenzoic acid

AC24595-0050 AC24595-0250
200009-150EA 105 USD
200009-150 200022-422
5-Nitroanthranilic acid 98%
5-Nitroanthranilic acid
Formula: C₇H₆N₂O₄
MW: 182.14 g/mol
Melting Pt: 278 °C
Storage Temperature: Ambient
MDL Number: MFCD00017039
CAS Number: 616-79-5
EINECS: 210-493-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR