(±)-3-Butyne-2-ol 97%

Danger

Synonyms: (±)-But-3-yn-2-ol(±)-3-Hydroxy-1-butyne

AC24531-0100 AC24531-0500
200029-220EA 34.9 USD
200029-220 200041-484
(±)-3-Butyne-2-ol 97%
(±)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 104…108 °C (1013 hPa)
Melting Pt: –12 °C
Density: 0.89 g/cm³
Flash Pt: 26 °C
MDL Number: MFCD00004541
CAS Number: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR