2,4-Dibromoanisole 98%

Synonyms: 2,4-Dibromo-1-methoxybenzene1,3-Dibromo-4-methoxybenzene

AC24323-0500 AC24323-0050
200040-622EA 12.2 USD
200040-622 200034-324
2,4-Dibromoanisole 98%
2,4-Dibromoanisole
Formula: C₇H₆Br₂O
MW: 265.93 g/mol
Boiling Pt: 101…106 °C (1 mmHg)
Melting Pt: 60…64 °C
MDL Number: MFCD00000079
CAS Number: 21702-84-1
EINECS: 244-536-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR