Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Difluoro-4-nitrobenzene ≥99%

Supplier: ACROS ORGANICS MS

Warning
Synonyms: 3,4-Difluoronitrobenzene

AC24193-1000 AC24193-0250
103531-954EA 175.43 USD InStock
103531-954 200029-074
1,2-Difluoro-4-nitrobenzene ≥99%
1,2-Difluoro-4-nitrobenzene


Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
Boiling Pt: 80°C - 81°C (14 mm Hg)
Density: 1.43 g/cm³ (20 °C)
Flash Pt: 80°C
Storage Temperature: Ambient
MDL Number: MFCD00007198
CAS Number: 369-34-6
EINECS: 206-718-2
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5080 to 1.5100 (20°C, 589 nm)
GC ≥98.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty