L(-)-Glucose 98%

Synonyms: (2S,3R,4S,5S)-2,3,4,5,6-Pentahydroxyhexanal

AC24192-0050 AC24192-2500 AC24192-0010
103511-822EA 307 USD
103511-822 200037-732 200019-432
L(-)-Glucose 98%
L(-)-Glucose
Formula: C₆H₁₂O₆
MW: 180.16 g/mol
Melting Pt: 153…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00148913
CAS Number: 921-60-8
EINECS: 213-068-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR