2,3-Dichlorobenzaldehyde 99%

Danger

AC23267-5000 AC23267-1000 AC23267-0250
200016-028EA 52.6 USD
200016-028 200040-536 103531-938
2,3-Dichlorobenzaldehyde 99%
2,3-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 144 °C (30 mmHg)
Melting Pt: 60…64 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00010127
CAS Number: 6334-18-5
EINECS: 228-711-3
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 64 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR