2,4-Difluorotoluene 98%

Danger

Synonyms: 2,4-Difluoro-1-methylbenzene

AC23221-0050 AC23221-0250
200022-194EA 151 USD
200022-194 200034-364
2,4-Difluorotoluene 98%
2,4-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 1.15 g/cm³
Flash Pt: 13 °C
MDL Number: MFCD00000332
CAS Number: 452-76-6
EINECS: 207-211-9
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4475 to 1.4495 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR