Fuchsin (acid), pure, high purity biological stain

Warning

Synonyms: Fuchsin SBenzenesulphonic acid, 2-amino-5-[(4-amino-3-sulphophenyl)(4-imino-3-sulpho-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)CN DcR-83-(1-(4-Amino-3-methyl-5-sulphonatophenyl)-1-(4-amino-3-sulphonatophenyl)methylene)cyclohexa-1,4-dienesulphonic acidAcid FuchsinAcid Fuchsin sodium saltRubin SAcid Magenta3-(1-(4-Amino-3-methyl-5-sulfonatophenyl)-1-(4-amino-3-sulfonatophenyl)methylene)cyclohexa-1,4-dienesulfonic acidAcid Violet 19CI 42685Benzenesulfonic acid, 2-amino-5-[(4-amino-3-sulfophenyl)(4-imino-3-sulfo-2,5-cyclohexadien-1-ylidene)methyl]-3-methyl-, sodium salt (1:2)

AC22790-1000
200036-456EA 195 USD
200036-456
Fuchsin (acid), pure, high purity biological stain
Fuchsin (acid)
Formula: C₂₀H₁₇N₃Na₂O₉S₃
MW: 585.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013286
CAS Number: 3244-88-0
EINECS: 221-816-5
Merck Index: 12,00109

Order Now

Specification Test Results

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
UV Lambda max 544 to 550 nm (in pH 3 acidified water)
Absorption Ratio P-15/P+15 1 to 1.4
Specific absorbance A (1%/1cm) ≥700 (at lambda max.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR