β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate 97+%

AC22757-1000 AC22757-0025 AC22757-5000
103467-022EA 636 USD
103467-022 102775-134 200002-772
β-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate 97+%
beta-Nicotinamide adenine dinucleotide phosphate sodium salt hydrate
Formula: C21H27N7NaO17P3 · xH2O
Melting Pt: 175…178 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00150480
CAS Number: 698999-85-8
EINECS: 214-664-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay ≥97.0 % (NADP, mono-Na) (calc. from enzymatic determination, based on dry weight)
Water ≤6.0 % (K.F.)
Ratio of Absorbances 0.80 to 0.86 (A250/A260, pH 7.5)
Ratio of Absorbances 0.18 to 0.24 (A280/A260, pH 7.5)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR